Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ppp1r14b
Entrez Gene ID
100759839
Gene Name
protein phosphatase 1, regulatory (inhibitor) subunit 14B
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615068: 289858 - 291781 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007651813.1 XP_007650003.1 1 - 2 196 94
GenBank mRNA JP050564.1 - 1 - 2 196 94

View Full Product Details