Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vwa2
Entrez Gene ID
100771172
Gene Name
von Willebrand factor A domain containing 2
Gene Aliases
I79_024267
Chromosome Location
Chr.Un|NW_003617093: 4423 - 11833 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007654937.1 XP_007653127.1 5 - 6 1417 102

View Full Product Details