Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Foxo1
Entrez Gene ID
100768647
Gene Name
forkhead box O1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614568: 159727 - 164819 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007649265.1 XP_007647455.1 1 - 2 1424 95

View Full Product Details