Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ddi1
Entrez Gene ID
100755779
Gene Name
DNA-damage inducible 1 homolog 1 (S. cerevisiae)
Gene Aliases
I79_016145
Chromosome Location
Chr.Un|NW_003614510: 504340 - 505545 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508260.1 XP_003508308.1 1 - 1 1126 104

View Full Product Details