Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Penk
Entrez Gene ID
100774423
Gene Name
proenkephalin
Gene Aliases
I79_021680
Chromosome Location
Chr.Un|NW_003615612: 188819 - 194947 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003512833.1 XP_003512881.1 1 - 2 117 93

View Full Product Details