Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc39a3
Entrez Gene ID
100774781
Gene Name
solute carrier family 39 (zinc transporter), member 3
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614244: 819413 - 824880 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007646990.1 XP_007645180.1 3 - 4 488 79

View Full Product Details