Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tti2
Entrez Gene ID
100760553
Gene Name
TELO2 interacting protein 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613666: 151834 - 161362 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003497839.2 XP_003497887.1 3 - 4 1315 98
GenBank mRNA JI890284.1 - 3 - 4 98

View Full Product Details