Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hspb2
Entrez Gene ID
100772331
Gene Name
heat shock 27kDa protein 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613694: 1131177 - 1132136 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003498527.2 XP_003498575.1 1 - 2 94 115

View Full Product Details