Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Htr6
Entrez Gene ID
100752934
Gene Name
5-hydroxytryptamine (serotonin) receptor 6, G protein-coupled
Gene Aliases
I79_014435
Chromosome Location
Chr.Un|NW_003614302: 412607 - 425877 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506892.2 XP_003506940.1 1 - 2 949 73

View Full Product Details