Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sox30
Entrez Gene ID
100763103
Gene Name
SRY (sex determining region Y)-box 30
Gene Aliases
I79_009450
Chromosome Location
Chr.Un|NW_003613914: 163330 - 210484 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007643426.1 XP_007641616.1 6 - 7 1367 121

View Full Product Details