Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klhl21
Entrez Gene ID
100760231
Gene Name
kelch-like family member 21
Gene Aliases
I79_018479
Chromosome Location
Chr.Un|NW_003614845: 30376 - 37065 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007650838.1 XP_007649028.1 1 - 2 342 77

View Full Product Details