Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ifi44l
Entrez Gene ID
100760542
Gene Name
interferon-induced protein 44-like
Gene Aliases
I79_001828
Chromosome Location
Chr.Un|NW_003613622: 1467522 - 1479658 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003496444.2 XP_003496492.2 4 - 5 830 93
XM_007650080.1 XP_007648270.1 4 - 5 830 93

View Full Product Details