Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam57a
Entrez Gene ID
100763695
Gene Name
family with sequence similarity 57, member A
Gene Aliases
I79_014190
Chromosome Location
Chr.Un|NW_003614276: 588615 - 617479 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007647260.1 XP_007645450.1 3 - 4 428 101
XM_007647261.1 XP_007645451.1 2 - 3 120 101
GenBank mRNA JI873577.1 - 1 - 2 101 101

View Full Product Details