Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Alx4
Entrez Gene ID
100750505
Gene Name
ALX homeobox 4
Gene Aliases
I79_015498
Chromosome Location
Chr.Un|NW_003614421: 749610 - 764131 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007648362.1 XP_007646552.1 2 - 3 470 62

View Full Product Details