Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Aatk
Entrez Gene ID
100753989
Gene Name
apoptosis-associated tyrosine kinase
Gene Aliases
I79_002370
Chromosome Location
Chr.Un|NW_003613637: 2280712 - 2313226 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007653957.1 XP_007652147.1 4 - 5 677 66
XM_007653960.1 XP_007652150.1 4 - 5 429 66
XM_007653965.1 XP_007652155.1 4 - 5 399 66

View Full Product Details