Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Efna4
Entrez Gene ID
100773413
Gene Name
ephrin-A4
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613854: 509754 - 513823 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501649.2 XP_003501697.1 2 - 3 362 63

View Full Product Details