Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kcnj3
Entrez Gene ID
100768536
Gene Name
potassium inwardly-rectifying channel, subfamily J, member 3
Gene Aliases
I79_012491
Chromosome Location
Chr.Un|NW_003614140: 217678 - 382598 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003505308.2 XP_003505356.1 1 - 2 700 97

View Full Product Details