Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ehd1
Entrez Gene ID
100761397
Gene Name
EH-domain containing 1
Gene Aliases
I79_019087
Chromosome Location
Chr.Un|NW_003614946: 253402 - 277679 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510724.2 XP_003510772.1 4 - 5 839 94
GenBank mRNA JP041866.1 - 3 - 4 629 94

View Full Product Details