Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prol1
Entrez Gene ID
100769334
Gene Name
proline rich, lacrimal 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615639: 149464 - 152044 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003512880.1 XP_003512928.1 1 - 2 52 88

View Full Product Details