Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mb
Entrez Gene ID
100762299
Gene Name
myoglobin
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003617782: 3351 - 11920 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003515322.2 XP_003515370.1 3 - 4 362 67

View Full Product Details