Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cyhr1
Entrez Gene ID
100755112
Gene Name
cysteine/histidine-rich 1
Gene Aliases
I79_015345
Chromosome Location
Chr.Un|NW_003614411: 123321 - 137720 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003507635.2 XP_003507683.1 2 - 3 676 80

View Full Product Details