Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pglyrp4
Entrez Gene ID
100771725
Gene Name
peptidoglycan recognition protein 4
Gene Aliases
I79_019771
Chromosome Location
Chr.Un|NW_003615070: 293640 - 306343 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003511291.1 XP_003511339.1 5 - 6 624 63

View Full Product Details