Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vstm2b
Entrez Gene ID
100771836
Gene Name
V-set and transmembrane domain containing 2B
Gene Aliases
I79_023080
Chromosome Location
Chr.Un|NW_003616254: 8771 - 13623 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003513979.1 XP_003514027.1 3 - 4 580 86

View Full Product Details