Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bud13
Entrez Gene ID
100773647
Gene Name
BUD13 homolog (S. cerevisiae)
Gene Aliases
BUD13-like
Chromosome Location
Chr.Un|NW_003615029: 332119 - 349227 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007651654.1 XP_007649844.1 2 - 3 776 77
GenBank mRNA JI868956.1 - 2 - 3 133 77
JP037249.1 - 2 - 3 133 77

View Full Product Details