Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Btd
Entrez Gene ID
100755324
Gene Name
biotinidase
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613589: 1210189 - 1241835 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003495343.2 XP_003495391.1 2 - 3 347 75

View Full Product Details