Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tnk1
Entrez Gene ID
100765201
Gene Name
tyrosine kinase, non-receptor, 1
Gene Aliases
I79_002762
Chromosome Location
Chr.Un|NW_003613643: 1908626 - 1914459 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003497186.2 XP_003497234.1 5 - 6 869 116
XM_007608450.1 XP_007606640.1 5 - 6 869 116

View Full Product Details