Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Siah2
Entrez Gene ID
100773133
Gene Name
siah E3 ubiquitin protein ligase 2
Gene Aliases
I79_009563
Chromosome Location
Chr.Un|NW_003613922: 620041 - 768571 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003502890.2 XP_003502938.1 3 - 4 226 103

View Full Product Details