Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Btnl2
Entrez Gene ID
100767924
Gene Name
butyrophilin-like 2 (MHC class II associated)
Gene Aliases
I79_024695
Chromosome Location
Chr.Un|NW_003616946: 10184 - 23649 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003515227.1 XP_003515275.1 2 - 3 423 104

View Full Product Details