Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trappc2l
Entrez Gene ID
100753676
Gene Name
trafficking protein particle complex 2-like
Gene Aliases
I79_000222
Chromosome Location
Chr.Un|NW_003613583: 916074 - 921178 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007609257.1 XP_007607447.1 1 - 2 80 89

View Full Product Details