Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Exph5
Entrez Gene ID
100753332
Gene Name
exophilin 5
Gene Aliases
I79_009409
Chromosome Location
Chr.Un|NW_003613912: 1441949 - 1509305 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007643379.1 XP_007641569.1 2 - 3 614 115

View Full Product Details