Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mal
Entrez Gene ID
100773093
Gene Name
mal, T-cell differentiation protein
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613665: 1697266 - 1720673 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003497801.1 XP_003497849.1 1 - 2 98 69
XM_007617928.1 XP_007616118.1 1 - 2 98 69

View Full Product Details