Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hoxd10
Entrez Gene ID
100752695
Gene Name
homeobox D10
Gene Aliases
I79_004131
Chromosome Location
Chr.Un|NW_003613688: 1451713 - 1454093 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003498317.2 XP_003498365.1 1 - 2 744 68

View Full Product Details