Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dusp9
Entrez Gene ID
100752633
Gene Name
dual specificity phosphatase 9
Gene Aliases
I79_013975
Chromosome Location
Chr.Un|NW_003614255: 593148 - 655073 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007647058.1 XP_007645248.1 3 - 4 590 59

View Full Product Details