Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Iqcf3
Entrez Gene ID
100767457
Gene Name
IQ motif containing F3
Gene Aliases
I79_006601
Chromosome Location
Chr.Un|NW_003613788: 239666 - 240951 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003500369.1 XP_003500417.1 2 - 3 159 99

View Full Product Details