Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Enpp7
Entrez Gene ID
100755684
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase 7
Gene Aliases
I79_014163
Chromosome Location
Chr.Un|NW_003614274: 522923 - 526470 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506698.1 XP_003506746.1 3 - 4 1017 71

View Full Product Details