Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ldlrad1
Entrez Gene ID
100771340
Gene Name
low density lipoprotein receptor class A domain containing 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613599: 3875320 - 3884006 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007643653.1 XP_007641843.1 3 - 4 201 80

View Full Product Details