Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Txlng
Entrez Gene ID
100752164
Gene Name
taxilin gamma
Gene Aliases
I79_017379
Chromosome Location
Chr.Un|NW_003614676: 396481 - 449974 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509272.2 XP_003509320.1 7 - 8 1108 110
GenBank mRNA JI891023.1 - 1 - 2 74 110
JP038792.1 - 1 - 2 74 110

View Full Product Details