Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mpv17l2
Entrez Gene ID
100770899
Gene Name
MPV17 mitochondrial membrane protein-like 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613716: 1042390 - 1043966 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007637836.1 XP_007636026.1 3 - 4 412 59

View Full Product Details