Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam8a1
Entrez Gene ID
100761518
Gene Name
family with sequence similarity 8, member A1
Gene Aliases
I79_001319
Chromosome Location
Chr.Un|NW_003613609: 1883917 - 1892572 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007645998.1 XP_007644188.1 1 - 2 135 126

View Full Product Details