Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Emx2
Entrez Gene ID
100750961
Gene Name
empty spiracles homeobox 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613779: 572102 - 679573 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007640900.1 XP_007639090.1 1 - 2 37 65

View Full Product Details