Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmcc3
Entrez Gene ID
100765845
Gene Name
transmembrane and coiled-coil domain family 3
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614514: 431690 - 587676 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007648912.1 XP_007647102.1 2 - 3 1032 73
XM_007648913.1 XP_007647103.1 2 - 3 1041 73
XM_007648914.1 XP_007647104.1 2 - 3 987 73
XM_007648915.1 XP_007647105.1 2 - 3 942 73

View Full Product Details