Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sult6b1
Entrez Gene ID
100757336
Gene Name
sulfotransferase family, cytosolic, 6B, member 1
Gene Aliases
I79_015615
Chromosome Location
Chr.Un|NW_003614440: 717159 - 734329 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003507861.1 XP_003507909.1 5 - 6 627 165

View Full Product Details