Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam46b
Entrez Gene ID
100771132
Gene Name
family with sequence similarity 46, member B
Gene Aliases
I79_016184
Chromosome Location
Chr.Un|NW_003614517: 184300 - 229535 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508303.2 XP_003508351.1 1 - 2 174 104

View Full Product Details