Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dlk1
Entrez Gene ID
100762551
Gene Name
delta-like 1 homolog (Drosophila)
Gene Aliases
I79_018083
Chromosome Location
Chr.Un|NW_003614787: 264502 - 272382 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509868.1 XP_003509916.1 1 - 2 39 83

View Full Product Details