Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Scaf1
Entrez Gene ID
100767426
Gene Name
SR-related CTD-associated factor 1
Gene Aliases
I79_023219
Chromosome Location
Chr.Un|NW_003616327: 74903 - 84718 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007654280.1 XP_007652470.1 5 - 6 516 56

View Full Product Details