Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mtif3
Entrez Gene ID
100763635
Gene Name
mitochondrial translational initiation factor 3
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613630: 2972624 - 2981825 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003496676.2 XP_003496724.1 2 - 3 499 98
XM_007652179.1 XP_007650369.1 3 - 4 603 98
GenBank mRNA JI874238.1 - 1 - 2 301 98
JP046981.1 - 1 - 2 301 98

View Full Product Details