Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fcf1
Entrez Gene ID
100771331
Gene Name
FCF1 rRNA-processing protein
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615144: 239328 - 249194 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003511529.2 XP_003511577.1 4 - 5 340 80
GenBank mRNA JP041294.1 - 1 - 2 117 80

View Full Product Details