Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wbscr28
Entrez Gene ID
100768388
Gene Name
Williams-Beuren syndrome chromosome region 28
Gene Aliases
I79_023626
Chromosome Location
Chr.Un|NW_003616582: 77089 - 81516 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003514409.1 XP_003514457.1 2 - 3 501 69

View Full Product Details