Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Creg1
Entrez Gene ID
100767565
Gene Name
cellular repressor of E1A-stimulated genes 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613889: 1025007 - 1037812 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003502285.2 XP_003502333.1 2 - 3 512 78
XM_007642893.1 XP_007641083.1 2 - 3 510 78
GenBank mRNA JP058209.1 - 2 - 3 169 78

View Full Product Details