Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cartpt
Entrez Gene ID
100757030
Gene Name
CART prepropeptide
Gene Aliases
I79_010562
Chromosome Location
Chr.Un|NW_003613989: 266997 - 268503 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003503682.2 XP_003503730.1 2 - 3 282 79
XM_003503683.2 XP_003503731.1 2 - 3 243 79

View Full Product Details